Product Details

SNP ID
rs2508343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85631973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAAAAAAAAGAATGGGGGAAAC[A/C]AGAACAGATGAAAATGGTGGTTTGT
Phenotype
MIM: 603583 MIM: 615533
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DLG2 PubMed Links

Gene Details

Gene
DLG2
Gene Name
discs large MAGUK scaffold protein 2
There are no transcripts associated with this gene.

Gene
TMEM126B
Gene Name
transmembrane protein 126B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193537.2 Intron NP_001180466.1
NM_001193538.2 Intron NP_001180467.1
NM_001256546.1 Intron NP_001243475.1
NM_001256547.1 Intron NP_001243476.1
NM_018480.4 Intron NP_060950.3
XM_011545165.2 Intron XP_011543467.1

View Full Product Details