Product Details

SNP ID
rs2683162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73750369 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAAGATGAACAGCTCCTGTCCC[C/T]GCGGTGGGCTGATGAGAGTGGAACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00469 PubMed Links

Gene Details

Gene
LINC00469
Gene Name
long intergenic non-protein coding RNA 469
There are no transcripts associated with this gene.

Gene
LOC100134391
Gene Name
uncharacterized LOC100134391
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278587.1 Intron NP_001265516.1
XM_017023987.1 Intron XP_016879476.1

View Full Product Details