Product Details

SNP ID
rs1124581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2906827 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGAACTCCTCCTCCGTCTCAGG[C/T]CGGGGCAGCAGCTTTCTGGGAGGAG
Phenotype
MIM: 607210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD11 PubMed Links

Gene Details

Gene
CARD11
Gene Name
caspase recruitment domain family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324281.1 3680 Silent Mutation CGA,CGG R1092R NP_001311210.1
NM_032415.5 3680 Silent Mutation CGA,CGG R1092R NP_115791.3
XM_011515586.2 3680 Silent Mutation CGA,CGG R1092R XP_011513888.1
XM_011515587.2 3680 Silent Mutation CGA,CGG R1091R XP_011513889.1

View Full Product Details