Product Details

SNP ID
rs2746024
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:18260834 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ATCGGGGGCGTGGCCCGCGATCACC[T/G]CTGGGGAAGGCGGGGAGGGGACCTT
Phenotype
MIM: 600362 MIM: 615498 MIM: 601243
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
FLII PubMed Links

Gene Details

Gene
FLII
Gene Name
FLII, actin remodeling protein
There are no transcripts associated with this gene.

Gene
MIEF2
Gene Name
mitochondrial elongation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144900.1 Intron NP_001138372.1
NM_139162.3 Intron NP_631901.2
NM_148886.1 Intron NP_683684.2
XM_017024190.1 Intron XP_016879679.1
Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
There are no transcripts associated with this gene.

View Full Product Details