Product Details
- SNP ID
-
rs2232569
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48568355 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAATTGCTACCAAGGAGCGGAGCG[A/G]GAGAGGCTTAATAACAGAAACCTGA
- Phenotype
-
MIM: 149750
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LALBA
PubMed Links
Gene Details
- Gene
- LALBA
- Gene Name
- lactalbumin alpha
View Full Product Details