Product Details

SNP ID
rs2233023
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:11675805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACTGGGCCCAGCCTCTTCCCAC[C/T]TCCCTTGCTGGCTCAGCAGCACCCC
Phenotype
MIM: 605647 MIM: 604094
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO6 PubMed Links

Gene Details

Gene
FBXO6
Gene Name
F-box protein 6
There are no transcripts associated with this gene.

Gene
MAD2L2
Gene Name
MAD2 mitotic arrest deficient-like 2 (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127325.1 Intron NP_001120797.1
NM_006341.3 Intron NP_006332.3
XM_011540507.1 Intron XP_011538809.1

View Full Product Details