Product Details

SNP ID
rs2233043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2250287 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAATCTGAGAACAGACCGTGGGC[A/G]GCTATGCTGACCAGGGCTCCGGATG
Phenotype
MIM: 613430 MIM: 610887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS3 PubMed Links

Gene Details

Gene
HAUS3
Gene Name
HAUS augmin like complex subunit 3
There are no transcripts associated with this gene.

Gene
MIR4800
Gene Name
microRNA 4800
There are no transcripts associated with this gene.

Gene
MXD4
Gene Name
MAX dimerization protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006454.2 919 UTR 3 NP_006445.1
XM_011513388.2 919 UTR 3 XP_011511690.1
XM_017007655.1 919 Intron XP_016863144.1
XM_017007656.1 919 Intron XP_016863145.1
Gene
POLN
Gene Name
DNA polymerase nu
There are no transcripts associated with this gene.

View Full Product Details