Product Details

SNP ID
rs2233363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49210110 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTTGCTAACACCCCTCTGGCAA[C/T]CTCTCTGGTCATGTCTTCCCGGTGT
Phenotype
MIM: 606363 MIM: 139391
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABI3 PubMed Links

Gene Details

Gene
ABI3
Gene Name
ABI family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135186.1 Intron NP_001128658.1
NM_016428.2 Intron NP_057512.1
XM_005257429.2 Intron XP_005257486.1
XM_011524873.1 Intron XP_011523175.1
XM_017024721.1 Intron XP_016880210.1
Gene
GNGT2
Gene Name
G protein subunit gamma transducin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198754.1 Intron NP_001185683.1
NM_001198755.1 Intron NP_001185684.1
NM_001198756.1 Intron NP_001185685.1
NM_031498.2 Intron NP_113686.1

View Full Product Details