Product Details

SNP ID
rs2233695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103776965 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGCCACAAAGCAGACTAGCCAGC[C/T]GGCTACAATTGGAGTCAGAGTCCCA
Phenotype
MIM: 300401
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLP1 PubMed Links

Gene Details

Gene
PLP1
Gene Name
proteolipid protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000533.4 173 UTR 5 NP_000524.3
NM_001128834.2 173 UTR 5 NP_001122306.1
NM_001305004.1 173 UTR 5 NP_001291933.1
NM_199478.2 173 UTR 5 NP_955772.1

View Full Product Details