Product Details

SNP ID
rs2233714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79347539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCGCCCGTGTCCCGCCCGACC[C/G]GCGCCCGCGATGCTGGCGCTGCGCT
Phenotype
MIM: 604486
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPIF PubMed Links

Gene Details

Gene
PPIF
Gene Name
peptidylprolyl isomerase F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005729.3 74 UTR 5 NP_005720.1
XM_005269379.2 74 UTR 5 XP_005269436.1

View Full Product Details