Product Details

SNP ID
rs2233818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:139454696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCACAGTAGCGTGTGTAAGCACA[A/G]TGGAAGTGAGCTGAGCAGAACCCAG
Phenotype
MIM: 180280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507291 PubMed Links

Gene Details

Gene
LOC100507291
Gene Name
uncharacterized LOC100507291
There are no transcripts associated with this gene.

Gene
RBP2
Gene Name
retinol binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004164.2 Intron NP_004155.2

View Full Product Details