Product Details

SNP ID
rs2233861
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:112406194 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATGGGCCTCAGACACTTGTGGCA[A/T]TAAGTGTCTACCATGTAGGCAGTAG
Phenotype
MIM: 603703
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RPL6 PubMed Links

Gene Details

Gene
RPL6
Gene Name
ribosomal protein L6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000970.4 Intron NP_000961.2
NM_001024662.2 Intron NP_001019833.1
NM_001320137.1 Intron NP_001307066.1
NM_001320138.1 Intron NP_001307067.1
NM_001320139.1 Intron NP_001307068.1
NM_001320140.1 Intron NP_001307069.1
NM_001320141.1 Intron NP_001307070.1
NM_001320142.1 Intron NP_001307071.1
XM_017019781.1 Intron XP_016875270.1
XM_017019782.1 Intron XP_016875271.1

View Full Product Details