Product Details

SNP ID
rs2073342
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:20892057 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTGCACAGAATATTTCAAACTGCA[C/G]GTATGCAGACAGACCAGGAAGGAGG
Phenotype
MIM: 131398
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNASE3 PubMed Links

Gene Details

Gene
RNASE3
Gene Name
ribonuclease A family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002935.2 425 Missense Mutation ACG,AGG T124R NP_002926.2

View Full Product Details