Product Details

SNP ID
rs2952785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:268030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGCTGTACATAAAGGCAGTGG[A/T]CCATGAGCCGATTTTCAGACCCCTG
Phenotype
MIM: 171500
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ACP1 PubMed Links

Gene Details

Gene
ACP1
Gene Name
acid phosphatase 1, soluble
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040649.2 Intron NP_001035739.1
NM_004300.3 Intron NP_004291.1
NM_007099.3 Intron NP_009030.1
Gene
SH3YL1
Gene Name
SH3 and SYLF domain containing 1
There are no transcripts associated with this gene.

View Full Product Details