Product Details

SNP ID
rs2235931
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:17023460 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GTTGCCTAGAGTCTGTGAAAAGCAC[A/G]TATCTAAAAAAGCACAAGCTTTCTG
Phenotype
MIM: 185470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDHB PubMed Links

Gene Details

Gene
SDHB
Gene Name
succinate dehydrogenase complex iron sulfur subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003000.2 Intron NP_002991.2

View Full Product Details