Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001301089.1 | 10186 | Intron | NP_001288018.1 | ||
NM_014758.2 | 10186 | Intron | NP_055573.2 | ||
XM_005271546.4 | 10186 | UTR 3 | XP_005271603.1 | ||
XM_011542819.2 | 10186 | UTR 3 | XP_011541121.1 | ||
XM_011542820.2 | 10186 | UTR 3 | XP_011541122.1 | ||
XM_011542821.2 | 10186 | UTR 3 | XP_011541123.1 | ||
XM_011542822.2 | 10186 | Intron | XP_011541124.1 | ||
XM_011542824.2 | 10186 | UTR 3 | XP_011541126.1 | ||
XM_011542825.2 | 10186 | UTR 3 | XP_011541127.1 | ||
XM_011542826.2 | 10186 | UTR 3 | XP_011541128.1 | ||
XM_011542827.2 | 10186 | UTR 3 | XP_011541129.1 | ||
XM_017017725.1 | 10186 | UTR 3 | XP_016873214.1 | ||
XM_017017726.1 | 10186 | UTR 3 | XP_016873215.1 |