Product Details

SNP ID
rs2270405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87498766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGACCTCGGGGTGCCGTGGTGCC[C/G]GGCCCCACGCGCGCGCGGGGCTGAG
Phenotype
MIM: 600831
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DAPK1 PubMed Links

Gene Details

Gene
DAPK1
Gene Name
death associated protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288729.1 Intron NP_001275658.1
NM_001288730.1 Intron NP_001275659.1
NM_001288731.1 Intron NP_001275660.1
NM_004938.3 Intron NP_004929.2
XM_005251757.3 Intron XP_005251814.1
XM_017014360.1 Intron XP_016869849.1

View Full Product Details