Product Details
- SNP ID
-
rs2270527
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:14735076 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTATTGAAGTTTGGGGAGAAAAGC[G/T]GGGTACATTCAGGTCCTTCTTGACG
- Phenotype
-
MIM: 608944
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FREM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117358199] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FREM1
- Gene Name
- FRAS1 related extracellular matrix 1
View Full Product Details