Product Details

SNP ID
rs2270919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58117039 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAACCACGGTAGCCCAAGAGGC[A/G]TACAAAGAAATTTGTGACACAGCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP20 PubMed Links

Gene Details

Gene
CFAP20
Gene Name
cilia and flagella associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013242.2 Intron NP_037374.1

View Full Product Details