Product Details

SNP ID
rs2974527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115124883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCACATTTAAAAATGTCTACAT[C/G]ATATGTGCTTGTACAAGGCTTGAGT
Phenotype
MIM: 609317
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRIM36 PubMed Links

Gene Details

Gene
TRIM36
Gene Name
tripartite motif containing 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017397.1 4081 Intron NP_001017397.1
NM_001017398.1 4081 Intron NP_001017398.1
NM_001300752.1 4081 UTR 3 NP_001287681.1
NM_001300759.1 4081 UTR 3 NP_001287688.1
NM_018700.3 4081 UTR 3 NP_061170.2
XM_017009621.1 4081 Intron XP_016865110.1
XM_017009622.1 4081 Intron XP_016865111.1
XM_017009623.1 4081 Intron XP_016865112.1

View Full Product Details