Product Details

SNP ID
rs2291754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18480471 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAATGATAAACTGCAATAACTTA[C/T]TCTGGGCACCTACTGATAAAAGGAA
Phenotype
MIM: 601387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDHAL6A PubMed Links

Gene Details

Gene
LDHAL6A
Gene Name
lactate dehydrogenase A like 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144071.1 1388 Intron NP_001137543.1
NM_144972.4 1388 Intron NP_659409.2
XM_005252805.4 1388 Intron XP_005252862.2
XM_011519922.2 1388 Intron XP_011518224.1
XM_011519923.2 1388 Intron XP_011518225.1
XM_011519924.2 1388 Intron XP_011518226.1
XM_011519925.1 1388 Intron XP_011518227.1
Gene
TSG101
Gene Name
tumor susceptibility 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006292.3 1388 UTR 3 NP_006283.1
XM_005253108.3 1388 UTR 3 XP_005253165.1

View Full Product Details