Product Details

SNP ID
rs2280708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123009703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGGAGCCGGAGCCGGTGCCGGT[C/T]CCTCTTCCATGAACTTAAGCTGAAA
Phenotype
MIM: 600301 MIM: 606238
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACADSB PubMed Links

Gene Details

Gene
ACADSB
Gene Name
acyl-CoA dehydrogenase, short/branched chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001609.3 Intron NP_001600.1
XM_017016194.1 Intron XP_016871683.1
Gene
IKZF5
Gene Name
IKAROS family zinc finger 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271840.1 Intron NP_001258769.1
XM_006717947.3 Intron XP_006718010.1
XM_017016550.1 Intron XP_016872039.1
XM_017016551.1 Intron XP_016872040.1
XM_017016552.1 Intron XP_016872041.1
XM_017016553.1 Intron XP_016872042.1
XM_017016554.1 Intron XP_016872043.1
XM_017016555.1 Intron XP_016872044.1

View Full Product Details