Product Details
- SNP ID
-
rs2999472
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:112393259 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACTACTATAGTTGATGACAGAAT[A/G]AGAACTAGACTCCAGATTTTTTAAC
- Phenotype
-
MIM: 615100
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTTNBP2NL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs530981832,rs550691766] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTTNBP2NL
- Gene Name
- CTTNBP2 N-terminal like
View Full Product Details