Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033925.1 | 3643 | UTR 3 | NP_001029097.1 | ||
NM_001323964.1 | 3643 | UTR 3 | NP_001310893.1 | ||
NM_001323965.1 | 3643 | UTR 3 | NP_001310894.1 | ||
NM_001323967.1 | 3643 | UTR 3 | NP_001310896.1 | ||
NM_001323968.1 | 3643 | UTR 3 | NP_001310897.1 | ||
NM_001323969.1 | 3643 | UTR 3 | NP_001310898.1 | ||
NM_001323970.1 | 3643 | UTR 3 | NP_001310899.1 | ||
NM_003252.3 | 3643 | UTR 3 | NP_003243.1 | ||
XM_017016605.1 | 3643 | Intron | XP_016872094.1 | ||
XM_017016606.1 | 3643 | UTR 3 | XP_016872095.1 |