Product Details

SNP ID
rs2305247
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:135170722 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGCAATGTCATAAAGTGTTGTAGGA[G/T]TTTCTTCACCCAGGCTTTGAGGATC
Phenotype
MIM: 602536 MIM: 615655
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB3GAP1 PubMed Links

Gene Details

Gene
RAB3GAP1
Gene Name
RAB3 GTPase activating protein catalytic subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172435.1 Intron NP_001165906.1
NM_012233.2 Intron NP_036365.1
XM_011510823.2 Intron XP_011509125.1
XM_011510825.2 Intron XP_011509127.1
Gene
ZRANB3
Gene Name
zinc finger RANBP2-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286568.1 Intron NP_001273497.1
NM_001286569.1 Intron NP_001273498.1
NM_032143.3 Intron NP_115519.2
XM_005263809.1 Intron XP_005263866.1
XM_006712788.1 Intron XP_006712851.1
XM_011511958.2 Intron XP_011510260.1
XM_011511959.2 Intron XP_011510261.2
XM_011511960.1 Intron XP_011510262.1
XM_011511961.2 Intron XP_011510263.2
XM_011511963.1 Intron XP_011510265.1
XM_011511966.2 Intron XP_011510268.2
XM_017005065.1 Intron XP_016860554.1
XM_017005066.1 Intron XP_016860555.1
XM_017005067.1 Intron XP_016860556.1
XM_017005068.1 Intron XP_016860557.1
XM_017005069.1 Intron XP_016860558.1
XM_017005070.1 Intron XP_016860559.1
XM_017005071.1 Intron XP_016860560.1

View Full Product Details