Product Details

SNP ID
rs3024977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57097467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCGAGTGTCATACTTCGGGAAG[A/C]AAAATGTCTAGGCCATTTCAAGGCC
Phenotype
MIM: 602381 MIM: 616496 MIM: 601512
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAB2 PubMed Links

Gene Details

Gene
NAB2
Gene Name
NGFI-A binding protein 2
There are no transcripts associated with this gene.

Gene
NEMP1
Gene Name
nuclear envelope integral membrane protein 1
There are no transcripts associated with this gene.

Gene
STAT6
Gene Name
signal transducer and activator of transcription 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178078.1 Intron NP_001171549.1
NM_001178079.1 Intron NP_001171550.1
NM_001178080.1 Intron NP_001171551.1
NM_001178081.1 Intron NP_001171552.1
NM_003153.4 Intron NP_003144.3
XM_006719575.2 Intron XP_006719638.1
XM_011538703.2 Intron XP_011537005.1
XM_011538704.2 Intron XP_011537006.1
XM_011538705.2 Intron XP_011537007.1
XM_011538706.2 Intron XP_011537008.1
XM_011538707.2 Intron XP_011537009.1
XM_011538708.2 Intron XP_011537010.1
XM_011538709.2 Intron XP_011537011.1

View Full Product Details