Product Details

SNP ID
rs2391191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:105467097 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATCTACTTCATAGGTTTTCAAA[A/G]GAGCATTCTTCTGAGCAAATCTGAA
Phenotype
MIM: 607408 MIM: 607415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAOA PubMed Links

Gene Details

Gene
DAOA
Gene Name
D-amino acid oxidase activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161812.1 208 UTR 5 NP_001155284.1
NM_001161814.1 208 UTR 5 NP_001155286.1
NM_172370.4 208 Missense Mutation AAG,AGG K30R NP_758958.3
XM_005254042.1 208 Missense Mutation AAG,AGG K30R XP_005254099.1
Gene
DAOA-AS1
Gene Name
DAOA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details