Product Details

SNP ID
rs2509401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119355222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCAACATCTGCCAACTGATTTC[G/T]CAAAGCTATTATTTATTCTGTACAA
Phenotype
MIM: 608752 MIM: 606227 MIM: 604725
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C1QTNF5 PubMed Links

Gene Details

Gene
C1QTNF5
Gene Name
C1q and tumor necrosis factor related protein 5
There are no transcripts associated with this gene.

Gene
MFRP
Gene Name
membrane frizzled-related protein
There are no transcripts associated with this gene.

Gene
USP2
Gene Name
ubiquitin specific peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243759.1 2687 UTR 3 NP_001230688.1
NM_004205.4 2687 UTR 3 NP_004196.4
NM_171997.2 2687 UTR 3 NP_741994.1
XM_005271721.4 2687 Intron XP_005271778.1
XM_005271722.2 2687 Intron XP_005271779.1
XM_017018539.1 2687 UTR 3 XP_016874028.1

View Full Product Details