Product Details

SNP ID
rs2512499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:87042906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGTGAATCTTTGTGTTTTCACA[A/G]TTTGTAAAGAATATCAGCTCTCACT
Phenotype
MIM: 616360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM135 PubMed Links

Gene Details

Gene
TMEM135
Gene Name
transmembrane protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168724.1 Intron NP_001162195.1
NM_022918.3 Intron NP_075069.3
XM_017018140.1 Intron XP_016873629.1
XM_017018141.1 Intron XP_016873630.1
XM_017018142.1 Intron XP_016873631.1

View Full Product Details