Product Details

SNP ID
rs2501801
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:17309868 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATGGGTGTGAAGGAAATGACAGG[A/G]GCTGAAGCTAGCCCAGGGCCTCTCC
Phenotype
MIM: 605347
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PADI4 PubMed Links

Gene Details

Gene
PADI4
Gene Name
peptidyl arginine deiminase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012387.2 Intron NP_036519.2
XM_011541150.1 Intron XP_011539452.1
XM_011541151.1 Intron XP_011539453.1
XM_011541152.1 Intron XP_011539454.1
XM_011541153.1 Intron XP_011539455.1
XM_011541154.2 Intron XP_011539456.1
XM_011541155.1 Intron XP_011539457.1
XM_011541156.1 Intron XP_011539458.1
XM_011541157.1 Intron XP_011539459.1

View Full Product Details