Product Details

SNP ID
rs2620633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144486673 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCCTCTTCAGTGAGGTGTGTCTT[C/T]GCATCCTCTGCCCATTTTCTAATTG
Phenotype
MIM: 609172
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R16A PubMed Links

Gene Details

Gene
PPP1R16A
Gene Name
protein phosphatase 1 regulatory subunit 16A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005272350.2 Intron XP_005272407.1
XM_011517349.2 Intron XP_011515651.1
XM_017013912.1 Intron XP_016869401.1
XM_017013913.1 Intron XP_016869402.1
XM_017013914.1 Intron XP_016869403.1
XM_017013915.1 Intron XP_016869404.1

View Full Product Details