Product Details

SNP ID
hCV16039829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:20326365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTCTGTTGAAAAAAACACAGAA[G/A]TTAAATAAAGCTCTATTCTATTATG
Phenotype
MIM: 608793
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CCDC144CP PubMed Links

Gene Details

Gene
CCDC144CP
Gene Name
coiled-coil domain containing 144C, pseudogene
There are no transcripts associated with this gene.

Gene
SPECC1
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
There are no transcripts associated with this gene.

View Full Product Details