Product Details
- SNP ID
-
rs2582871
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:21994334 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCCTTCCCAGCTGTATGGCAGATA[A/G]GTTTGGCTCTGCCAATACCAAGAAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRAMENP
PubMed Links
Gene Details
- Gene
- PRAMENP
- Gene Name
- PRAME N-terminal-like, pseudogene
There are no transcripts associated with this gene.
View Full Product Details