Product Details

SNP ID
rs2589787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:38260201 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACCTTCAAGGATTTCAGTGAAGG[A/G]CTTCTTTCTAACTTTCCTCCTTAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGFLAM PubMed Links

Gene Details

Gene
EGFLAM
Gene Name
EGF like, fibronectin type III and laminin G domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205301.1 Intron NP_001192230.1
NM_152403.3 Intron NP_689616.2
NM_182798.2 Intron NP_877950.1
NM_182801.2 Intron NP_877953.1

View Full Product Details