Product Details
- SNP ID
-
rs2609433
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70637479 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGCATGGTGTGTGTTTGTTGGG[G/T]GTTGTGGAGGGTATGTTTTTGAGTG
- Phenotype
-
MIM: 606585
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ENAM
PubMed Links
Gene Details
- Gene
- ENAM
- Gene Name
- enamelin
View Full Product Details