Product Details

SNP ID
rs2687956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39455810 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTTAAATTTTTTTTAAAGCTAC[C/T]GTGTTTACTATGTATTACGCATGCA
Phenotype
MIM: 611135 MIM: 607031 MIM: 603686
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLB PubMed Links

Gene Details

Gene
KLB
Gene Name
klotho beta
There are no transcripts associated with this gene.

Gene
LIAS
Gene Name
lipoic acid synthetase
There are no transcripts associated with this gene.

Gene
RPL9
Gene Name
ribosomal protein L9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000661.4 Intron NP_000652.2
NM_001024921.3 Intron NP_001020092.1

View Full Product Details