Product Details

SNP ID
rs2695321
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:39623891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGGCAAGAAATGAGGCTGGAA[A/G]TGAGGTTGGGACCAAAAGAAGACCT
Phenotype
MIM: 609034
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEYL PubMed Links

Gene Details

Gene
HEYL
Gene Name
hes related family bHLH transcription factor with YRPW motif-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014571.3 3654 UTR 3 NP_055386.1
XM_005270745.2 3654 UTR 3 XP_005270802.1

View Full Product Details