Product Details
- SNP ID
-
rs2741161
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52675153 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCCAAGCATATTTGTTAGTGATG[A/C]TGGGGGAGAACTAGAGCTAATGAAA
- Phenotype
-
MIM: 139350
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT1
PubMed Links
Gene Details
- Gene
- KRT1
- Gene Name
- keratin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006121.3 |
2034 |
UTR 3 |
|
|
NP_006112.3 |
View Full Product Details