Product Details

SNP ID
rs2769031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107065843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATTGTTTTATACAACTGTGTTT[C/T]ACATTAGTTAAGAGAAAAAAGAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM41 PubMed Links

Gene Details

Gene
RBM41
Gene Name
RNA binding motif protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171080.1 3052 Intron NP_001164551.1
NM_001324242.1 3052 UTR 3 NP_001311171.1
NM_001324243.1 3052 UTR 3 NP_001311172.1
NM_001324244.1 3052 Intron NP_001311173.1
NM_018301.4 3052 UTR 3 NP_060771.3
XM_011530980.2 3052 Intron XP_011529282.1
XM_011530981.2 3052 UTR 3 XP_011529283.1

View Full Product Details