Product Details

SNP ID
rs2753281
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:86869239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTAAACCTGCAAGTACACTTCTA[A/G]GACTTTATTCTGAGGAAATAACTTA
Phenotype
MIM: 606254
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEP15 PubMed Links

Gene Details

Gene
SEP15
Gene Name
15 kDa selenoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004261.3 Intron NP_004252.2
NM_203341.1 Intron NP_976086.1

View Full Product Details