Product Details

SNP ID
rs2734732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4883300 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAATGCTTGGCCTGCACCAGGGC[A/T]AGGGAGAGGACGACACCAAGGAGGT
Phenotype
MIM: 602871 MIM: 609771
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PPL PubMed Links

Gene Details

Gene
PPL
Gene Name
periplakin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002705.4 5457 UTR 3 NP_002696.3
XM_006720902.3 5457 UTR 3 XP_006720965.1
XM_017023374.1 5457 UTR 3 XP_016878863.1
XM_017023375.1 5457 UTR 3 XP_016878864.1
Gene
UBN1
Gene Name
ubinuclein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079514.2 5457 Intron NP_001072982.1
NM_001288656.1 5457 Intron NP_001275585.1
XM_005255277.3 5457 Intron XP_005255334.1
XM_005255278.3 5457 Intron XP_005255335.1
XM_005255279.3 5457 Intron XP_005255336.1
XM_011522465.1 5457 Intron XP_011520767.1
XM_011522466.2 5457 Intron XP_011520768.1
XM_011522467.1 5457 Intron XP_011520769.1
XM_017023189.1 5457 Intron XP_016878678.1
XM_017023190.1 5457 Intron XP_016878679.1
XM_017023191.1 5457 Intron XP_016878680.1
XM_017023192.1 5457 Intron XP_016878681.1

View Full Product Details