Product Details

SNP ID
rs2854737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44374691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTCGGGGCAGGCTGAGCGGGG[C/G]CACCGCATAGGGGAGGGAGGACACG
Phenotype
MIM: 607759
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITGA2B PubMed Links

Gene Details

Gene
ITGA2B
Gene Name
integrin subunit alpha 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000419.4 3039 Missense Mutation CCC,GCC P971A NP_000410.2
XM_011524749.1 3039 Intron XP_011523051.1
XM_011524750.1 3039 Missense Mutation CCC,GCC P971A XP_011523052.1

View Full Product Details