Product Details

SNP ID
rs2855455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33165154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTGCAGCATCCTACTGCTGCAGC[C/T]CACTTTCATCACGTGACACCTCTGC
Phenotype
MIM: 120290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL11A2 PubMed Links

Gene Details

Gene
COL11A2
Gene Name
collagen type XI alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163771.1 Intron NP_001157243.1
NM_080679.2 Intron NP_542410.2
NM_080680.2 Intron NP_542411.2
NM_080681.2 Intron NP_542412.2
XM_011514299.2 Intron XP_011512601.1
XM_011514300.2 Intron XP_011512602.1
XM_011514302.2 Intron XP_011512604.1
XM_017010250.1 Intron XP_016865739.1
XM_017010251.1 Intron XP_016865740.1

View Full Product Details