Product Details

SNP ID
rs2131048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74938448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAGGGAAGGCCGCAGAGAGGAGGC[A/G]GGGTCTCTGCCGAGTCCTGAAGGAC
Phenotype
MIM: 607827 MIM: 607828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTOP2 PubMed Links

Gene Details

Gene
OTOP2
Gene Name
otopetrin 2
There are no transcripts associated with this gene.

Gene
OTOP3
Gene Name
otopetrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272005.1 Intron NP_001258934.1
NM_178233.2 Intron NP_839947.1
XM_011524744.2 Intron XP_011523046.1

View Full Product Details