Product Details

SNP ID
rs2856758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46370170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATAGATTATATCTGGAGTGAAG[A/G]ATCCTGCCACCTATGTATCTGGCAT
Phenotype
MIM: 601267 MIM: 601373
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCR2 PubMed Links

Gene Details

Gene
CCR2
Gene Name
C-C motif chemokine receptor 2
There are no transcripts associated with this gene.

Gene
CCR5
Gene Name
C-C motif chemokine receptor 5 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000579.3 29 UTR 5 NP_000570.1
NM_001100168.1 29 UTR 5 NP_001093638.1
Gene
LOC102724297
Gene Name
uncharacterized LOC102724297
There are no transcripts associated with this gene.

View Full Product Details