Product Details

SNP ID
rs2894565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2389046 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCCACGGGAGTCCTCATCCTCAC[A/G]CAGCCGCCCAGCCAGCAGTCACGTT
Phenotype
MIM: 610477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS9 PubMed Links

Gene Details

Gene
TMPRSS9
Gene Name
transmembrane protease, serine 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527978.2 Intron XP_011526280.1
XM_011527980.1 Intron XP_011526282.1
XM_011527982.2 Intron XP_011526284.1

View Full Product Details