Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001001551.3 | 166 | UTR 5 | NP_001001551.2 | ||
NM_001256915.1 | 166 | UTR 5 | NP_001243844.1 | ||
XM_006717110.3 | 166 | Intron | XP_006717173.1 | ||
XM_006717111.3 | 166 | Intron | XP_006717174.1 | ||
XM_017014719.1 | 166 | Missense Mutation | CAT,TAT | H51Y | XP_016870208.1 |
XM_017014720.1 | 166 | Missense Mutation | CAT,TAT | H51Y | XP_016870209.1 |
XM_017014721.1 | 166 | Missense Mutation | CAT,TAT | H135Y | XP_016870210.1 |
XM_017014722.1 | 166 | Missense Mutation | CAT,TAT | H135Y | XP_016870211.1 |
XM_017014723.1 | 166 | Missense Mutation | CAT,TAT | H135Y | XP_016870212.1 |
XM_017014724.1 | 166 | Intron | XP_016870213.1 | ||
XM_017014725.1 | 166 | UTR 5 | XP_016870214.1 |