Product Details

SNP ID
rs2098070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:11602494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGTGGCCTCCTTGTTCCTCC[G/T]TGCTGCTCTAGATCCACATCAAAAA
Phenotype
MIM: 603330
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAH9 PubMed Links

Gene Details

Gene
DNAH9
Gene Name
dynein axonemal heavy chain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001372.3 Intron NP_001363.2
NM_004662.2 Intron NP_004653.2
XM_011523703.1 Intron XP_011522005.1
XM_017024292.1 Intron XP_016879781.1
XM_017024293.1 Intron XP_016879782.1
XM_017024294.1 Intron XP_016879783.1
XM_017024295.1 Intron XP_016879784.1

View Full Product Details