Product Details

SNP ID
rs2814161
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132190197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGTGTTTTTGCTGCCGTTGGTC[C/T]CCCCTCAGTGGGAGGCAGCCTCCTG
Phenotype
MIM: 608407 MIM: 611198
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DPYSL4 PubMed Links

Gene Details

Gene
DPYSL4
Gene Name
dihydropyrimidinase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006426.2 Intron NP_006417.2
XM_005252658.3 Intron XP_005252715.1
XM_011539125.2 Intron XP_011537427.1
XM_017015482.1 Intron XP_016870971.1
XM_017015483.1 Intron XP_016870972.1
Gene
JAKMIP3
Gene Name
Janus kinase and microtubule interacting protein 3
There are no transcripts associated with this gene.

View Full Product Details