Product Details

SNP ID
rs2099504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:6977616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTAGGACATTTGACACCCCCC[C/A]CCTATCACCTTTTTAAAACATTTCC
Phenotype
MIM: 400033
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
TBL1Y PubMed Links

Gene Details

Gene
TBL1Y
Gene Name
transducin (beta)-like 1, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033284.1 Intron NP_150600.1
NM_134258.1 Intron NP_599020.1
NM_134259.1 Intron NP_599021.1
XM_005262572.3 Intron XP_005262629.1
XM_017030086.1 Intron XP_016885575.1
XM_017030087.1 Intron XP_016885576.1

View Full Product Details